Learn More
Medchemexpress LLC HY-112786 1mg Medchemexpress, MC-Val-Cit-PAB-MMAF CAS:863971-17-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1127861MG
Encompass_Preferred
Medchemexpress, HY-112786 1mg MC-Val-Cit-PAB-MMAF CAS:863971-17-9 MC-Val-Cit-PAB-MMAF is a drug-linker conjugate for ADC with antitumor activity by using the tubulin inhibitor, MMAF, linked via cathepsin cleavable MC-Val-Cit-PAB. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9742