Learn More
Medchemexpress LLC MC-Val-Cit-PAB-MMAF | 863971-17-9 | 99.25% | 1330.61 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112786
Encompass_Preferred
MC-Val-Cit-PAB-MMAF (Vc-MMAF) is a drug-linker conjugate for ADC, utilizing the tubulin inhibitor, MMAF (HY-15579), linked via cathepsin cleavable MC-Val-Cit-PAB (HY-78738). It exhibits antitumor activity.
  • Drug-linker conjugate for ADC.
  • Uses tubulin inhibitor MMAF.
  • Linked via cathepsin cleavable MC-Val-Cit-PAB.
  • Shows antitumor activity.
Catalog No. 50-004-39412