Learn More
Medchemexpress LLC MC-Val-Cit-PAB-MMAF | 863971-17-9 | 99.3% | 1330.61 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11278650MG
Encompass_Preferred
MC-Val-Cit-PAB-MMAF (Vc-MMAF) is a drug-linker conjugate for Antibody-Drug Conjugate (ADC) applications. It utilizes the tubulin inhibitor MMAF, linked via a cathepsin cleavable MC-Val-Cit-PAB. This conjugate has demonstrated antitumor activity.
  • Used in antibody-drug conjugate (ADC) applications.
  • Utilizes the tubulin inhibitor MMAF.
  • Linked via a cathepsin cleavable MC-Val-Cit-PAB.
  • Demonstrates antitumor activity.
Catalog No. 50-004-38517