Learn More
Cayman Chemical Mc-Val-CIt-PABC-PNP 10mg
SDP
Supplier:  Cayman Chemical 2388110 MG
Encompass_Preferred
A peptide linker molecule; used in the synthesis of antibody-drug conjugates
Catalog No. 50-299-4466