Learn More
Apexbio Technology LLC Mc-Val-Cit-PABC-PNP 159857-81-5 10mg
SDP
Supplier:  Apexbio Technology LLC B100310
Encompass_Preferred
Mc-Val-Cit-PABC-PNP (159857-81-5) is a peptide-based linker commonly utilized in the development of antibody-drug conjugates (ADCs) It is designed to facilitate site-specific payload release through protease-mediated cleavage especially by lysosomal cathepsin enzymes enabling targeted drug delivery within tumor cells Mc-Val-Cit-PABC-PNP exerts its biological function by being recognized and cleaved upon internalization in cancer cells resulting in the selective release of conjugated cytotoxic agents Based on these properties Mc-Val-Cit-PABC-PNP holds research potential in the synthesis and optimization of ADCs for targeted cancer therapies
Catalog No. 50-000-90545