Learn More
Selleck Chemical LLC MC-VC-PAB-PNP
SDP
Supplier:  Selleck Chemical LLC S68325MG
Encompass_Preferred
MC-Val-Cit-PAB-PNP (Mc-Val-Cit-PABC-PNP) is a cathepsin cleavable ADC peptide linker that is applicable to the synthesis of antibody-drug conjugates (ADCs)
Catalog No. 50-000-02733