Learn More
Medchemexpress LLC MC-VC(S)-PABQ-Tubulysin M, 10mg
SDP
Supplier:  Medchemexpress LLC HY12891010MG
Encompass
MC-VC(S)-PABQ-Tubulysin M is a synthetic ADC drug-linker conjugate composed of the tubulin polymerization inhibitor Tubulysin M (an ADC Cytotoxin) (HY-N7053) and MC-VC(S)- PABQ (an ADC linker) is connected. MC-VC(S)-PABQ-Tubulysin M is effective against multidrug-resistant lymphoma cell lines and tumors.
Catalog No. NC3806259