Learn More
ATCC MC116; B. lymphocyte; Human (Homo sapiens)
Supplier:  ATCC CRL1649
Encompass
MC116 is a B lymphocyte cell that was isolated from the ascites of a lymphoma male patient. This cell line was deposited by IT Magrath and can be used in immunology research.
Catalog No. 50-238-1893
 Restricted Item