Learn More
Selleck Chemical LLC MC1568 10mg 852475-26-4
SDP
Supplier:  Selleck Chemical LLC S148410MG
Encompass_Preferred
MC1568 is a selective HDAC inhibitor for maize HD1-A with IC50 of 100 nM in a cell-free assay. It is 34-fold more selective for HD1-A than HD1-B. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-875-7