Learn More
Medchemexpress LLC HY-119940 10mg Medchemexpress, MC180295 CAS:2237942-08-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11994010MG
Encompass_Preferred
Medchemexpress, HY-119940 10mg MC180295 CAS:2237942-08-2 MC180295 ((rel)-MC180295) is a potent and selective CDK9-Cyclin T1 inhibitor, with an IC50 of 5 nM, at least 22-fold more selective for CDK9 over other CDKs. MC180295 also inhibits GSK-3α and GSK-3β. MC180295 ((rel)-MC180295) has potent anti-tumor effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9559