Learn More
Selleck Chemical LLC MC180295 5mg 2237942-08-2
SDP
Supplier:  Selleck Chemical LLC S88095MG
Encompass_Preferred
MC180295 is a novel potent and selective CDK9 inhibitor with an IC50 of 5 nM and is at least 22-fold more selective for CDK9 over other CDKs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1752170