Learn More
Medchemexpress LLC MC2590 | 2284460-01-9 | C20H17N3O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1522265MG
Encompass_Preferred
MC2590 is a potent pyridine-containing histone deacetylase (HDAC) inhibitor, with a purity of 98.71%. It effectively inhibits a broad spectrum of HDAC isoforms, leading to G2/M cell cycle arrest and modulation of microRNAs towards apoptosis induction.
  • Potent pyridine-containing HDAC inhibitor
  • Inhibits HDAC1-3, -6, -8, and -10 (class I/IIb-selective)
  • Also inhibits HDAC4, HDAC5, HDAC7, HDAC9, and HDAC11
  • Induces G2/M cell cycle arrest
  • Modulates pro- and anti-apoptotic microRNAs for apoptosis induction
Catalog No. 50-003-73205