Learn More
CPC Scientific MCA-Val-Asn-Leu-Asp-Ala-Glu-Phe-Lys(DNP)-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD106A
Encompass
ONE-LETTER SEQUENCE: Mca-VNLDAEF-Lys(Dnp)MOLECULAR FORMULA: C60H76N12O22MOLECULAR WEIGHT:1317.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: Alzheimer's Disease
Catalog No. 50-210-8728