Learn More
CPC Scientific MCA-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Dap(DNP)-OH (acetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD104A
Encompass
SEQUENCE: MCA-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Dap(DNP)-OH (acetate salt)ONE-LETTER SEQUENCE: Mca-SEVNLDAE-Dap(Dnp)MOLECULAR FORMULA: C56H73N13O26MOLECULAR WEIGHT: 1344.27STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: MCA-SER-GLU-VAL-ASN-LEU-ASP-ALA-GLU-DAP(DNP)MCA-SER-GLU-VAL-ASN-LEU-ASP-ALA-GLU-DAP(DNP)-OHMCA-(ASN670,LEU671)-AMYLOID BETA/A4 PROTEIN PRECURSOR770 (667-674)-DAP(DNP)MCA-(ASN670,LEU671)-APP770 (667-674)-DAP(DNP)Mca-Asn670,Leu671-Amyloid b/A4 Protein Precursor770: 667-674-Dap: DnpRESEARCH AREA: Alzheimer's DiseaseREFERENCES:SKU(s): AMYD-104A, AMYD-104B
Catalog No. 50-210-8724