Learn More
Carbosynth LLC. MOCAc-Lys-Pro-Leu-Gly-Leu-Dap(Dnp)-Ala-Arg-NH2, 5mg, MMP-1 and MMP-2 substrate, MW: 1221.35, Molecular formula: C55H80N16O16, CAS# 720710-69-0
SDP
Supplier:  Carbosynth LLC. SMO3670PI5MG
Encompass_Preferred
Synonyms: Mca-Lys-Pro-Leu-Gly-Leu-Dap(Dnp)-Ala-Arg-NH2, Mca-KPLGL-Dap(Dnp)-AR-NH2, Application: Fluorescence-Quenching Substrate for MMPs, Cathepsin D and E, ADAM10, and ADAM17/TACE, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C, References: U. Neumann , H. Kubota, K. Frei, V. Ganu, and D. Leppert, Anal. Biochem., 328, 166 (2004).
Catalog No. 50-168-7136