Learn More
Medchemexpress LLC Mca-Pro-Leu-Gly-Pro-D-Lys(Dnp) | 185698-23-1 | 99.3% | 892.91 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP37921MG
Encompass_Preferred
Mca-Pro-Leu-Gly-Pro-D-Lys(Dnp) is a FRET substrate of Thimet oligopeptidase. This product can be utilized for the determination of Thimet oligopeptidase activity.
  • FRET substrate of Thimet oligopeptidase.
  • Can be used for the determination of Thimet oligopeptidase activity.
Catalog No. 50-004-00214