Learn More
Apexbio Technology LLC Mca-Pro-Leu-NH2 5mg
SDP
Supplier:  Apexbio Technology LLC A15015
Encompass_Preferred
Mca-Pro-Leu-NH2 is a synthetic dipeptide substrate labeled with a fluorescent 7-methoxycoumarinacetyl (Mca) group Upon cleavage by specific proteases or peptidases such as aminopeptidases the Mca fluorophore is released generating a measurable fluorescence signal that correlates with enzyme activity Mca-Pro-Leu-NH2 is utilized for real-time monitoring of proteolytic reactions enzyme kinetic studies protease inhibitor screening and pharmacological research Its enzyme-dependent fluorescence emission makes it valuable for protease characterization inhibitor profiling and drug discovery experiments
Catalog No. 50-000-96641