Learn More
Medchemexpress LLC Mca-Pro-Leu-OH | 140430-55-3 | 99.1% | 444.48 | 5 MG

Supplier: Medchemexpress LLC HYP46455MG
Mca-Pro-Leu-OH is a fluorescently labeled dipeptide with a molecular formula of C23H28N2O7. It appears as a white to off-white solid and is intended for research use only.
- Fluorescently labeled dipeptide
- Molecular formula: C23H28N2O7
- Appearance: solid, white to off-white
- Sequence: {Mca-Pro}-Leu
- Shipped at room temperature in continental US
- Recommended storage for powder: -80°C for 2 years, -20°C for 1 year
- Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
- Store sealed, away from moisture and light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.