Learn More
Cayman Chemical McaSEVNLDAEFRK DnpRRNH2 5mg
SDP
Supplier:  Cayman Chemical 368795 MG
Encompass_Preferred
A fluorogenic substrate for BACE; Mca is released upon cleavage by BACE and its fluorescence can be used to quantify BACE activity; ex/em = 328/420 nm, respectively
Catalog No. 50-291-5192