Learn More
Medchemexpress LLC MCB-22-174 | 3058199-58-6 | 99.8% | 429.37 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY165607S
Encompass_Preferred
MCB-22-174 is a deuterated Piezo1 agonist with an EC50 value of 6.28 μM. It activates the CaMKII/ERK signaling pathway and initiates Ca2+ influx in rMSCs. The compound can improve bone quality in hind-limb unloading (HU) model rats and can be used for the study of disuse osteoporosis (OP). It also decreases the expression of chondrogenesis markers (Comp, Acan) and adipogenesis markers (Lpl, Fabp4) in MSCs.
  • Activates CaMKII/ERK signaling pathway
  • Initiates Ca2+ influx in rMSCs
  • Decreases expression of chondrogenesis markers (Comp, Acan)
  • Decreases expression of adipogenesis markers (Lpl, Fabp4)
  • Improves bone quality in hind-limb unloading model rats
  • Useful for disuse osteoporosis (OP) studies
Catalog No. 50-003-95154