Learn More
Medchemexpress LLC MCC | 104676-09-7 | 95.0% | 236.27 g/mol | C12H16N2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY13225110MG
Encompass_Preferred
MCC is a non-cleavable antibody-drug conjugate (ADC) linker used in the synthesis of ADCs such as MCC-DM1. It is supplied for research use only and is provided in multiple pack sizes with reported purity of ≥95.0%, molecular weight 236.27 g/mol, and formula C12H16N2O3. Manufacturer-provided documents include a data sheet, safety data sheet (SDS), certificate of analysis (COA), H NMR, and mass spectrometry (MS) files.
  • Non-cleavable ADC linker for conjugation chemistry
  • Purity ≥95.0%
  • Available in multiple small-scale pack sizes (5 mg-500 mg)
  • Manufacturer documents: data sheet, SDS, COA, H NMR, MS
  • For research use only; not for human use
Catalog No. 50-002-03792