Learn More
Medchemexpress LLC MCC | 104676-09-7 | 95.0% | 236.27 g·mol⁻¹ | C12H16N2O3 | 50 MG

Supplier: Medchemexpress LLC HY13225150MG
MCC is a non-cleavable antibody-drug conjugate (ADC) linker used to attach cytotoxic payloads to antibodies for targeted delivery. Supplied for research use only, it is provided as a solid and should be stored under inert atmosphere for long-term stability.
- Non-cleavable ADC linker suitable for ADC synthesis
- Purity ≥95.0%
- Molecular weight 236.27 g·mol⁻¹
- Chemical formula C12H16N2O3
- Appearance: solid, light yellow to brown
- Storage: store under nitrogen; in solvent: -80°C (up to 6 months), -20°C (1 month)
- Intended for research use only; not for human use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.