Learn More
Selleck Chemical LLC MCC950 Sodium
SDP
Supplier:  Selleck Chemical LLC S78091G
Encompass_Preferred
MCC950 Sodium is a potent selective inhibitor of NLRP3 with IC50 of 7 5 nM in BMDMs but not the AIM2 NLRC4 or NLRP1 inflammasomes
Catalog No. 50-000-01154