Learn More
Cayman Chemical MCC950sodIum salt 5mg
SDP
Supplier:  Cayman Chemical 347625 MG
Encompass_Preferred
An inhibitor of NLRP3 inflammasome activation; inhibits ATP-induced IL-1β release in LPS-primed mouse BMDMs (IC50 = 7.5 nM), as well as cytosolic LPS-induced IL-1β release in Pam3CSK4-primed mouse BMDMs at 0.1 and 1 µM; selective for NLRP3 over NLRC4 and AIM2 inflammasomes and does not inhibit LPS-induced NLRP3 priming in mouse BMDMs at 10 µM; reduces ox-LDL-induced increases in caspase-1 activity and inhibits pyroptosis in THP-1 macrophages at 1 µM; reduces myocardial fibrosis in mice following myocardial infarction induced by left coronary artery ligation at 10 mg/kg; improves forelimb grip strength and reduces spinal edema in a mouse model of spinal crush injury at 10 mg/kg
Catalog No. 50-291-2250