Learn More
Alkali Scientific McCown's Woody Plant Medium (Powder), 10 L
SDP
Supplier:  Alkali Scientific MCC002CP
Encompass_Preferred
McCown's Woody Plant Medium is provided in powder form to make 10 liters of nutrient solution. This formulation is specifically designed to support the growth of woody plant species in vitro, making it a valuable tool in plant tissue culture for research, forestry, and horticulture applications. The medium contains essential nutrients, vitamins, and minerals that help woody plants thrive in a laboratory environment, and is widely used for plant propagation, somatic embryogenesis, and genetic modification. It is ideal for the culture of both hardwood and softwood species and is used to optimize growth and regeneration in woody plant research.
Catalog No. 50-006-61665