Learn More
Abcam mCherry ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2218291X96TESTS
Encompass_Preferred
mCherry ELISA Kit is a single-wash 90-min SimpleStep Sandwich ELISA used to quantify mCherry with a sensitivity of 0.47 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric assay - 450 nm readout, works on any plate reader - Validated in cell culture extracts, cell culture media and cell culture supernatant - Need to design your own assay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1907493