Learn More
Nanocomposix NanoXact Mesoporous Silica Nanospheres - Aminated (Dried) - 100 nm, 25 mg
SDP
Supplier:  Nanocomposix SHAD10025MG
Encompass_Preferred
As a global leader in silica nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.100 nm spherical mesoporous silica nanoparticles with hexagonal MCM-41 pore structure combine biocompatibility and high drug-loading capacity with versatile surface functionalization capabilities. These nanoparticles are useful in a broad range of applications from nanomedicine, targeted drug delivery, catalysis, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, UV-Vis, and BET (pore size & surface area) characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. 50-195-9467