Learn More
ATCC MCM Genomic DNA; mock community; genomic material
Supplier:  ATCC MSA4000
Encompass
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product comprises genomic DNA prepared from fully sequenced, characterized, and authenticated ATCC Genuine Cultures, and was quantitated using Droplet Digital™ PCR to improve precision in relative abundance among strains within the community. Each order for this product is provided with access to run 20 analyses on One Codex, the leading bioinformatics platform for microbial genomics and metagenomics. Specification Range: 0.80 ng/uL to 1.33 ng/uL* *DNA concentrations indicate ATCC manufacturing specifications and are provided as a reference only
Catalog No. 50-238-2323