Learn More
TARGETMOL CHEMICALS INC MCN-A 343 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2296310MG
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. McN-A 343 is a selective M1 muscarinic agonist known for its ability to stimulate muscarinic transmission in sympathetic ganglia. It has been found to effectively reduce inflammation and oxidative stress in experimental models of ulcerative colitis. purity: 99%
Catalog No. 50-258-0907