Learn More
Aviva Systems Biology Corp MCOLN3 Antibody - middle region (ARP37698_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37698_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MCOLN3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MCOLN3. Alternate names: FLJ11006, FLJ36629, MGC71509, TRP-ML3, TRPML3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-685