Learn More
Medchemexpress LLC MCOPPB trihydrochloride | 1108147-88-1 | 100.0% | C26H43Cl3N4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1310110MG
Encompass_Preferred
MCOPPB trihydrochloride is a research-grade nociceptin/orphanin FQ (NOP) receptor agonist supplied as the trihydrochloride salt. It is a white to off-white solid with high reported purity, intended for in vitro and in vivo pharmacology studies targeting opioid-related receptors.
  • High reported purity suitable for research use.
  • Trihydrochloride salt form for enhanced stability and solubility control.
  • Available in multiple solid and solution pack sizes to support assay workflows.
  • Suitable for both in vitro biochemical assays and in vivo pharmacology models.
  • Accompanied by batch-specific COA documentation for quality assurance.
Catalog No. 50-002-60497