Learn More
Medchemexpress LLC MCPA-d3 | 352431-14-2 | 98.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0859S11MG
Encompass_Preferred
MCPA-d3 is a deuterium-labeled variant of MCPA, which is an orally active phenoxyacetic acid herbicide. This compound disrupts membrane integrity, energy metabolism by decreasing ATP levels, and the redox balance within plant cells. It also elevates hepatic cytochrome P-450 levels and boosts the activities of aniline hydroxylase and 7-ethoxycoumarin O-deethylase. It is effective in managing broadleaf weeds.
  • Can be utilized as a tracer.
  • Can serve as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08075