Learn More
Grainger MCR SAFETY Coated Gloves, M, 8
Supplier:  Grainger 33M007
Encompass_Preferred
The MCR Safety Coated Gloves M-8 are a pair of coated work gloves that provide protection, grip, and dexterity for general purpose tasks. They are made from a 13 gauge nylon shell with a sandy nitrile coated palm and fingers to resist oils, greases, and abrasions. The gloves have an ergonomic fit with keystone thumb design for reduced hand fatigue. They meet ANSI Level 2 cut resistance standards. The gloves are machine washable, making them reusable. They come in sizes S-2XL.
Catalog No. NC2207092