Learn More
Enterprise Technology Solutions MCR Safety B96699 Work Gloves, 13-Gauge Polyester, PU Coating, 12 Pairs/CT, Medium. Black
SDP
Supplier:  Enterprise Technology Solutions MCSCRWB96699M
Encompass_Preferred
The B96699 Work Gloves offer excellent comfort, grip and fit with good dexterity and abrasion-resistance. They are made with a comfortable and lightweight, 13-gauge black polyester fabric. Black polyurethane-coated palm and fingertips add abrasion-resistance and grip to make your workday better. Black-colored fabric and coating also hide dirt and grime. Color-coded hem identifies the medium size.
Catalog No. 50-008-28038