Learn More
Polycarbin Reversible PCR and Microcentrifuge Tube Rack, 80 wells
SDP
Supplier:  Polycarbin 2PR80
Encompass_Preferred
Polycarbin's reversible PCR tube and microcentrifuge tube racks offer an eco-friendly solution for benchtop work and freezer storage. Made from 100% recycled content, these durable, double-sided polypropylene racks accommodate 0.2 mL thin-walled PCR tubes on one side and 1.5 mL to 2 mL microcentrifuge tubes on the opposite side. Each side features 60 wells arranged in a 8 x 10 array, keeping tubes securely upright. Compact and designed for sustainability, these racks provide an efficient, reliable alternative for tube organization in any laboratory setting.
Catalog No. NC3683376