Learn More
Polycarbin Reversible PCR and Microcentrifuge Tube Rack, 96 wells
SDP
Supplier:  Polycarbin 2PR96
Encompass_Preferred
Polycarbin’s reversible PCR and microcentrifuge tube rack is an eco-friendly solution for benchtop work and freezer storage. Made from 100% recycled content, this durable, autoclavable polypropylene rack features a double-sided design, with one side accommodating 0.2 mL thin-walled PCR tubes and the opposite side holding 1.5–2.0 mL microcentrifuge tubes. Each side contains 96 wells arranged in a standard 8 x 12 array, ensuring a secure fit. Designed for sustainability and efficiency, this rack provides a reliable alternative for organizing and stabilizing tubes in any laboratory setting.
Catalog No. NC3683377