Learn More
Medchemexpress LLC MD-222 | 2136246-72-3 | 98.0% | 893.83 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY134823
Encompass_Preferred
MD-222 is the first-in-class highly potent PROTAC degrader of MDM2. It consists of ligands for Cereblon and MDM2. MD-222 induces rapid degradation of the MDM2 protein and activation of wild-type p53 in cells, exhibiting anticancer effects.
  • First-in-class highly potent PROTAC degrader of MDM2.
  • Consists of ligands for Cereblon and MDM2.
  • Induces rapid degradation of the MDM2 protein.
  • Activates wild-type p53 in cells.
  • Exhibits anticancer effects.
Catalog No. 50-003-65641