Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MD-224 1g | 2136247-12-4 | 1 G
SDP

Supplier:  Medchemexpress LLC HY1143121G

Encompass_Preferred

MD-224 is a PROTAC small molecule degrader that targets MDM2 for proteasomal degradation and is supplied for research use in cellular and biochemical studies of MDM2-p53 pathways and anticancer mechanisms.

  • Targets MDM2 for proteasomal degradation
  • Suitable for cellular and biochemical assays
  • Molecular formula C48H43Cl2FN6O6; molecular weight 889.80 g/mol
  • CAS number 2136247-12-4
  • Available in 1 G pack

Catalog No. 50-002-29162


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.