Learn More
Viva Products MD1000 Xpress Micro Dialyzer
SDP
Supplier:  Viva Products SCIMD1000DK0608
Encompass
MD1000 Xpress Mini Dialyzer Kit; For purification and desalting of proteins, antibodies, DNA and RNA; Convenient 48 well plate format to allow easy automated liquid handling; Sample volume up to 1.0 ml; Regenerated cellulose membrane; 6-8 kDa MWCO
Catalog No. NC1951752