Learn More
Dojindo Molecular Technologies Inc MDA(malondialdehyde) Assay Kit, 100test, Capable of measuring MDA in cells and tissues, Simple procedure.
SDP
Supplier:  Dojindo Molecular Technologies Inc M49610
Encompass
Malondialdehyde (MDA) is one of the end products of lipid peroxides, the MDA concentration in cell or tissue samples can indicate lipid peroxidation which is important in the fields of oxidative stress and ferroptosis. MDA is also called reactive aldehyde and reacts with amino and thiol groups to cause protein denaturation and DNA damage, so it is also measured in various disease studies such as cancer and diabetes. Dojindo's MDA Assay Kit uses the TBARS method to detect MDA by measuring the fluorescence or absorbance of the adduct of MDA and thiobarbituric acid that develops color according to the amount of MDA.
Catalog No. NC3441174