Learn More
ATCC MDA PCa 2b; Prostate Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL2422
Encompass
MDA PCa 2b is a cell line exhibiting epithelial-like morphology that was isolated from the prostate of a 63-year-old, Black male patient with adenocarcinoma. This cell line was deposited by NM Navone.
Catalog No. 50-238-3308
 Restricted Item