Learn More
Cayman Chemical MDA19 N 5hydroxyhexyl met 1mg
SDP
Supplier:  Cayman Chemical 360391 MG
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a potential metabolite of MDA 19 based on the published metabolism of JWH 018; intended for research and forensic applications
Catalog No. 50-291-2637