Learn More
Apexbio Technology LLC MDL-800, 5mg CAS# 2275619-53-7
SDP
Supplier:  Apexbio Technology LLC B83845
Encompass_Preferred
MDL-800 is a selective activator of SIRT6 which increases SIRT6 deacetylation activity by up to 22-fold, causing a decrease in H3K9Ac and H3K56Ac levels in human HCC cell, leads to cell cycle arrest and inhibition of cell proliferation. It also suppresses xenograft tumor growth HCC cells in immunocompromised mice. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8262