Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MDM2-IN-1 | 1410737-09-5 | 98.0% | 50 MG
SDP

Supplier:  Medchemexpress LLC HY13068450MG

Encompass_Preferred

MDM2-IN-1 is a synthetic MDM2-p53 interaction inhibitor with a trans-configuration. This compound is primarily utilized for the study of osteosarcoma, targeting the MDM-2/p53 pathway involved in apoptosis. It is supplied as a high-purity solid, with specific storage recommendations to maintain its stability and potency.

  • Synthetic MDM2-p53 interaction inhibitor
  • Features a trans-configuration
  • Targets MDM-2/p53 pathway
  • Used for osteosarcoma research
  • Purity of 98.0%
  • Supplied as a solid
  • Recommended solid storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Shipping at room temperature if less than 2 weeks

Catalog No. 50-003-40260


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.