Learn More
TARGETMOL CHEMICALS INC MDR-652 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T223605MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MDR-652 is a highly specific and efficacious agonist of nonpungent transient receptor potential vanilloid 1 (TRPV1), with Ki values of 11.4 nM and 23.8 nM for hTRPV1 and rTRPV1, respectively, and EC50s of 5.05 nM and 93 nM for hTRPV1 and rTRPV1, respectively. MDR-652 is a potent topical analgesic. Purity 99.96%
Catalog No. 50-287-7407