Learn More
Selleck Chemical LLC Enzalutamide (MDV3100) 10mg 915087-33-1
SDP
Supplier:  Selleck Chemical LLC S125010MG
Encompass_Preferred
Enzalutamide (MDV3100) is an androgen-receptor (AR) antagonist with IC50 of 36 nM in LNCaP cells. Enzalutamide is shown to increase autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-701-7