Learn More
Selleck Chemical LLC ME-344 S9736-5MG
SDP
Supplier:  Selleck Chemical LLC S97365MG
Encompass_Preferred
ME-344 is a synthetic metabolite of NV-128 a potent inhibitor of mitochondrial oxidative phosphorylation (OXPHOS) complex I disrupting ATP generation and oxygen consumption It also inhibits mTOR by downregulating the AKT/mTOR pathway exhibiting anticancer activity by inhibiting cell growth and viability in leukemia cell lines with IC50 values of 70 260 nM
Catalog No. 50-000-24909