Learn More
Medchemexpress LLC Me-CO-Lys(Cbo)-Gly-Arg-pNA | 96559-87-4 | 98.9% | 671.70 | 5 MG

Supplier: Medchemexpress LLC HYP47725MG
Me-CO-Lys(Cbo)-Gly-Arg-pNA is a substrate for C1 esterase, intended for research use only. This chemical compound is provided as an off-white to light yellow solid with a high purity.
- Purity: 98.9%
- Molecular weight: 671.70
- CAS number: 96559-87-4
- Powder storage: -80°C for 2 years, -20°C for 1 year (sealed storage, away from moisture)
- In solvent storage: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.