Learn More
Medchemexpress LLC ME1111 50mg | 1391758-52-3 | 50MG
SDP
Supplier:  Medchemexpress LLC HY10801250MG
Encompass_Preferred
ME1111 is an antifungal agent that is active against dermatophytes ME1111 is an inhibitor of the succinate dehydrogenase of Trichophyton species ME1111 has an excellent ability to penetrate human nails and is used for onychomycosis research[1 [2
Catalog No. 50-002-42125