Learn More
Aviva Systems Biology Corp ME3 Antibody - N-terminal region (ARP48511_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48511_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ME3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ME3. Alternate names: FLJ34862, NADP-MEApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59845